missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fructosyl amino acid oxidase Polyclonal specifically detects Fructosyl amino acid oxidase in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications
Specifications
| Antigen | Fructosyl amino acid oxidase |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot 1:500 - 1:5000, ELISA 1:5000 - 1:20000, Immunoprecipitation 1:100 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide |
| Gene Accession No. | Q8RIU8 |
| Gene Symbols | faoxC |
| Host Species | Sheep |
| Immunogen | Fructosyl-Amino Acid Oxidase [from Corynebacterium sp. Expressed in E.coli] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?