missing translation for 'onlineSavingsMsg'
Learn More

Fructosyl amino acid oxidase Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18411037 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411037 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411037 Supplier Novus Biologicals Supplier No. NBP177782AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Fructosyl amino acid oxidase Polyclonal antibody specifically detects Fructosyl amino acid oxidase in Bacteria samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fructosyl amino acid oxidase
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC=1.5.3.-, FAOX-C
Host Species Sheep
Immunogen Fructosyl amino acid oxidase [from Corynebacterium sp. Expressed in E.coli] (Uniprot: Q8RIU8)
Purification Method Multi-step
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.