missing translation for 'onlineSavingsMsg'
Learn More

Fructose-tryptophan (mixture of diastereomers), TRC

Product Code. 30394824
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394824 50 mg -
30399152 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394824 Supplier Toronto Research Chemicals Supplier No. TRCF79258050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25020-15-9
Molecular Formula C17H22N2O7
Molecular Weight (g/mol) 366.37
Synonym N-(1-Deoxy-D-fructos-1-yl)-L-tryptophan, (S)-1-[[1-Carboxy-2-(1H-indol-3-yl)ethyl]amino]-1-deoxy-D-fructose, D-Fructose-tryptophan, Fructose-L-tryptophan
SMILES OC1(CN[C@@H](CC2=CNC3=C2C=CC=C3)C(O)=O)[C@@H](O)[C@H](O)[C@H](O)CO1

Specifications

InChI Formula InChI=1S/C17H22N2O7/c20-8-14(22)16(24)15(23)13(21)7-19-12(17(25)26)5-9-6-18-11-4-2-1-3-10(9)11/h1-4,6,12,14-16,18-20,22-24H,5,7-8H2,(H,25,26)/t12-,14+,15+,16+/m0/s1
Quantity 50 mg
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Fructose-tryptophan (Mixture of Diastereomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.