missing translation for 'onlineSavingsMsg'
Learn More

Fructose, MedChemExpress

Product Code. 16441398
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441398 5 g 5g
16422529 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16441398 Supplier Medchem Express Supplier No. HYN0395
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fructose is a simple ketonic monosaccharide found in many plants, where it is often bonded to glucose to form the disaccharide sucrose.

Fructose is a simple ketonic monosaccharide found in many plants, where it is often bonded to glucose to form the disaccharide sucrose.

Specifications

Chemical Name or Material Fructose
CAS 7660-25-5
Color White
Purity Grade Notes Research
Molecular Formula C6H12O6
Quantity 5 g
Solubility Information H2O : 100 mg/mL (555.06 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (555.06 mM)
SMILES O[C@H]1[C@@H](O)[C@H](O)[C@@](O)(CO)OC1
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.