missing translation for 'onlineSavingsMsg'
Learn More

Fructose-asparagine TFA Salt(Mixture of diastereomers), TRC

Product Code. 30400983
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400983 10 mg
30406092 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400983 Supplier Toronto Research Chemicals Supplier No. TRCF79252710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10H18N2O8 • x(C2HF3O2)
Molecular Weight (g/mol) 408.28
Synonym N2-(1-Deoxy-D-fructos-1-yl)-L-asparagine TFA Salt, (S)-1-[(3-Amino-1-carboxy-3-oxopropyl)amino]-1-deoxy-D-fructose TFA Salt, 1-[(2-Carbamoyl-1-carboxyethyl)amino]-1-deoxy-fructose TFA Salt, 1-Deoxy-1-L-asparagino-D-fructose TFA Salt, N-(1-Deoxy-D-fructosyl)-L-asparagine TFA Salt
IUPAC Name ((3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexyl)-L-asparagine 2,2,2-trifluoroacetic acid
SMILES O=C(N)C[C@@H](C(O)=O)NCC([C@H]([C@@H]([C@@H](CO)O)O)O)=O.OC(C(F)(F)F)=O

Specifications

InChI Formula InChI=1S/C10H18N2O8.C2HF3O2/c11-7(16)1-4(10(19)20)12-2-5(14)8(17)9(18)6(15)3-13;3-2(4,5)1(6)7/h4,6,8-9,12-13,15,17-18H,1-3H2,(H2,11,16)(H,19,20);(H,6,7)/t4-,6+,8+,9+;/m0./s1
Quantity 10 mg
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Fructose-asparagine TFA Salt(Mixture of diastereomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.