missing translation for 'onlineSavingsMsg'
Learn More

FRS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7204343 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16819853 20 μL 20µL
16809853 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16819853 Supplier Proteintech Supplier No. 122491AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FRS3 (fibroblast growth factor receptor substrate 3), also known as FRS2B (FRS2-beta), is a 492 amino acid lipid-anchor adapter protein that contains one IRS-type PTB domain. Colocalizing to neural tissues with Tuj1, FRS3 functions as a feedback inhibitor of EGFR family members by preventing heterodimer formation between EGFR and ErbB2, thereby acting as a potential tumor suppressor. FRS3 is phosphorylated upon stimulation by FGF-2 or NGF and, acting as an adapter protein, links c-Fgr and NGF receptors to downstream signaling pathways. Interfering with the phosphorylation and nuclear translocation of ERK-2, FRS3 down-regulates ERK-2 expression. FRS3 likely interacts directly with GRB2, SH-PTP2, Flg, and Trk A, and may be involved in MAP kinase activation.

Specifications

Antigen FRS3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FRS3
Gene Accession No. O43559, Q52RG8, Q91WJ0
Gene Alias FGFR signaling adaptor SNT2, FGFR substrate 3, FRS2B, FRS2beta, FRS3, SNT 2, SNT2
Gene Symbols Frs3
Host Species Rabbit
Immunogen FRS3 Fusion Protein Ag2896
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107971, 10817, 316213
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.