missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536403
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 54 kDa. Purity is >95% by SDS-PAGE.
FRS3 (fibroblast growth factor receptor substrate 3), also known as FRS2B (FRS2-beta), is a 492 amino acid lipid-anchor adapter protein that contains one IRS-type PTB domain. Colocalizing to neural tissues with Tuj1, FRS3 functions as a feedback inhibitor of EGFR family members by preventing heterodimer formation between EGFR and ErbB2, thereby acting as a potential tumor suppressor. FRS3 is phosphorylated upon stimulation by FGF-2 or NGF and, acting as an adapter protein, links c-Fgr and NGF receptors to downstream signaling pathways. Interfering with the phosphorylation and nuclear translocation of ERK-2, FRS3 down-regulates ERK-2 expression. FRS3 likely interacts directly with GRB2, SH-PTP2, Flg, and Trk A, and may be involved in MAP kinase activation.
Specifications
| FRS3 | |
| Polyclonal | |
| Unconjugated | |
| FRS3 | |
| 4930417B13Rik; AI449674; FGFR substrate 3; FGFR-signaling adaptor SNT2; Fibroblast growth factor receptor substrate 3; FRS2B; Frs2beta; FRS2-beta; FRS3; Snt2; SNT-2; Suc1-associated neurotrophic factor target 2; suc1-associated neurotrophic factor target 2 (FGFR signalling adaptor); testicular tissue protein Li 71 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 107971, 10817, 316213 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43559, Q52RG8, Q91WJ0 | |
| FRS3 | |
| Synthetic peptide corresponding to amino acids 191-240 of human FRS3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction