missing translation for 'onlineSavingsMsg'
Learn More

FRS2 Antibody (SR2015), Novus Biologicals™

Rabbit Monoclonal Antibody

£257.00 - £341.00

Specifications

Antigen FRS2
Clone SR2015
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:20, Immunoprecipitation 1:50
Applications Western Blot, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18626664
View Documents
Novus Biologicals
NBP3-21759-100ul
100 μg
£341.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18642684
View Documents
Novus Biologicals
NBP3-21759-50ul
50 μg
£257.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FRS2 Monoclonal antibody specifically detects FRS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications

Specifications

FRS2
Western Blot 1:500-1:2000, Flow Cytometry 1:20, Immunoprecipitation 1:50
Monoclonal
Purified
RUO
Human
FGFR signalling adaptor, FGFR-signaling adaptor SNT, fibroblast growth factor receptor substrate 2, FRS2A, FRS2alpha, SNT, SNT1, SNT-1FGFR substrate 2, Suc1-associated neurotrophic factor target 1
A synthesized peptide derived from human FRS2 (Uniprot #: Q8WU20)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2015
Western Blot, Flow Cytometry, Immunoprecipitation
Unconjugated
Rabbit
Cancer, Signal Transduction
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
10818
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.