missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEAL5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568349
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
TCEAL5, which is located on the X chromosome, encodes a protein which contains a BEX (brain expressed X-liked like family) domain. This domain is found in proteins encoded by the TCEAL elongation factor (transcription elongation factor A (SII)-like) gene family also located on the X chromosome. The coding region for this gene is located entirely in the terminal exon.
Specifications
| TCEAL5 | |
| Polyclonal | |
| Unconjugated | |
| TCEAL5 | |
| TCEAL5 | |
| Synthesized peptide derived from human TCEAL5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5H9L2, Q8CCT4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 331532, 340543, 680282 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction