Learn More
OHAUS™ Frontier™ 5000 Multi-Pro Series Screw Cap Bucket
Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
83041011
Unit Size:
Pack of 2
Description
Frontier™ 5000 Multi-Pro Series centrifuge accessory.
- Screw cap
- 200 mL bucket
- 2 pack
Specifications
Specifications
| Type | Bucket |
| Material | Polypropylene |
| For Use With (Equipment) | 83041011 |
| Disposable | Reusable |
| Capacity | 200 mL |
| Autoclavable | Not Autoclavable |
| No. per Pack | 2 Pieces |