Learn More
OHAUS™ Frontier™ 5000 Multi-Pro Series Bucket
Shop All Ohaus Products
Click to view available options
Container Type:
With Cap
Without Cap
Unit Size:
Pack of 2
Description
Frontier™ Multi-Pro Series centrifuge accessory.
- Bucket
- 200 mL
- 2 pack
Specifications
Specifications
| For Use With (Equipment) | 83041011 |
| Capacity | 1 x 200 mL |
| Type | Bucket |
| Container Type | Without Cap |
| Material | Aluminum |
| Tube Size | 200 mL |