missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRMPD2 Polyclonal Antibody

FRMPD2 Polyclonal Antibody
Brand: Invitrogen PA580645
This item is not returnable.
View return policy
Description
FRMPD2 Polyclonal Antibody for IHC (P)
Specifications
| FRMPD2 | |
| Polyclonal | |
| Unconjugated | |
| FRMPD2 | |
| ENSMUSG00000071536; FERM and PDZ domain containing 2; FERM and PDZ domain-containing protein 2; FRMPD2; Gm1582; Gm626; PDZ domain containing 5C; PDZ domain-containing protein 4; PDZ domain-containing protein 5C; PDZD5C; PDZK4; PDZK5C; predicted gene 626; RGD1565652 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 143162 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q68DX3 | |
| FRMPD2 | |
| A synthetic peptide corresponding to the C-terminus of the Human FRMPD2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction