missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRMD6 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£174.00 - £393.00
Specifications
| Antigen | FRMD6 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18321826
|
Novus Biologicals
NBP3-05019-20UL |
20 μL |
£174.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18327854
|
Novus Biologicals
NBP3-05019-100UL |
100 μL |
£393.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FRMD6 Polyclonal specifically detects FRMD6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| FRMD6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 122786 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| FERM domain containing 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 405-614 of human FRMD6 (NP_689543.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title