Learn More
Invitrogen™ FRMD6 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5121253
Product Code. 17712654
  • £430.00 / 100µL
Description
Positive test controls include: SH-SY5Y, HeLa, HepG2, MCF7, Mouse kidney, OVCAR8, Rat lung, Rat spleen. The target is usually found in the following locations: Cell membrane, Cytoplasm, Cytoplasmic side, Peripheral membrane protein. Immunogen sequence: KEWKKEASKV RQYEVTWGID QFGPPMIIHF RVQYYVENGR LISDRAARYY YYWHLRKQVL HSQCVLREEA YFLLAAFALQ ADLGNFKRNK HYGKYFEPEA YF.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FRMD6
Polyclonal
Unconjugated
Frmd6
2610019M19Rik; 4.1 ezrin radixin moesin (FERM)-containing protein; 4930488L10Rik; AW212977; c14_5320; C14orf31; EX1; expanded homolog; FERM domain containing 6; FERM domain containing 6; FERM domain-containing protein 6; FERM domain-containing protein 163SCII; FERM domain-containing protein 6; FERM-domain-containing protein 163SCII; Frmd6; MGC17921; Willin
Rabbit
Affinity Chromatography
RUO
122786, 257646, 319710
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.95 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C0V9, Q8VII0, Q96NE9
Frmd6
A synthetic peptide.
100 μL
Primary
Human, Mouse, Rat