missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRMD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552850
This item is not returnable.
View return policy
Description
Immunogen sequence: SCPSITHGPR LSSVPRTRRR AVHISIMEGL ESLRDSAHST PVRSTSHGDT FLPHVRSSRT DSNERVAVIA DEAYSPADSV LPTPVAEHSL ELMLLSRQIN GATCSIEEEK ESEAST Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
FRMD5 is a protein coding gene.
Specifications
| FRMD5 | |
| Polyclonal | |
| Unconjugated | |
| FRMD5 | |
| 1500032A09Rik; 5830442F04; A930004K21Rik; FERM domain containing 5; FERM domain-containing protein 5; FRMD5; im:7161681; R75430; RGD1561554 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84978 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z6J6 | |
| FRMD5 | |
| Recombinant protein corresponding to Human FRMD5. Recombinant protein control fragment (Product #RP-90472). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction