missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRK Monoclonal Antibody (1A8H2C12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515425
This item is not returnable.
View return policy
Description
MA5-15425 targets FRK in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15425 immunogen is purified recombinant fragment of FRK (aa2-300) expressed in E. Coli.
FRK (PTK5) is a nuclear protein and may function during G1 and S phase of the cell cycle, suppressing growth. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.
Specifications
| FRK | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| P42685 | |
| FRK | |
| Purified recombinant fragment of FRK (aa2-300) expressed in E. Coli. | |
| RUO | |
| 2444 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1A8H2C12 | |
| Unconjugated | |
| FRK | |
| B-cell src-homology tyrosine kinase; beta-cell Src-homology tyrosine kinase; BSK; BSK/IYK; C85044; EC 2.7.1.112; EC 2.7.10.2; FRK; fyn related Src family tyrosine kinase; fyn-related kinase; fyn-related Src family tyrosine kinase; GASK; Gastrointestinal-associated kinase; gastrointestinal-associated tyrosine kinase; GTK; intestine tyrosine kinase; Iyk; Nuclear tyrosine protein kinase RAK; protein-tyrosine kinase 5; PTK5; PTK5 protein tyrosine kinase 5; RAK; src related tyrosine kinase; tyrosine-protein kinase FRK | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction