missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled 7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16974-1-AP-150UL
This item is not returnable.
View return policy
Description
Members of the ′frizzled′ gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD7 protein contains an N-terminal signal sequence, 10 cysteine residues typical of the cysteine-rich extracellular domain of Fz family members, 7 putative transmembrane domains, and an intracellular C-terminal tail with a PDZ domain-binding motif. FZD7 gene expression may downregulate APC function and enhance beta-catenin-mediated signals in poorly differentiated human esophageal carcinomas.Specifications
| Frizzled 7 | |
| Polyclonal | |
| Unconjugated | |
| FZD7 | |
| Frizzled 7, Fz 7, FZD7, FzE3, hFz7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100360552, 14369, 8324 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75084, Q61090 | |
| Fzd7 | |
| Frizzled 7 Fusion Protein Ag9446 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction