missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Frizzled-7 Polyclonal antibody specifically detects Frizzled-7 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Frizzled-7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Fz-7, FzE3frizzled-7, hFz7 |
| Host Species | Rabbit |
| Immunogen | Reacts with residues 1-12 [MRGPGTAASHSPC] of mouse fz7. 91% identity rat. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?