missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-02320
This item is not returnable.
View return policy
Description
Frizzled-7 Polyclonal antibody specifically detects Frizzled-7 in Human, Mouse, Rat, Bat, Bovine, Canine, Hamster, Primate, Monkey, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Frizzled-7 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Fz-7, FzE3frizzled-7, hFz7 | |
| Synthetic 18 amino acid peptide from C-terminus of human Frizzled-7. | |
| 0.05 mg | |
| GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
| 8324 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| ELISA, Immunohistochemistry 15 μg/mL, Immunohistochemistry-Paraffin 15 μg/mL | |
| O75084 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction