missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £365.00
Specifications
| Antigen | Frizzled-6 |
|---|---|
| Concentration | 1 mg/ml |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18335874
|
Novus Biologicals
NBP3-18335-0.025MG |
0.025 mg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18334576
|
Novus Biologicals
NBP3-18335-0.1MG |
0.1 mg |
£365.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Frizzled-6 Polyclonal antibody specifically detects Frizzled-6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
| Frizzled-6 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| frizzled (Drosophila) homolog 6, frizzled homolog 6 (Drosophila), frizzled-6, fz-6, FZ6, HFZ6, seven transmembrane helix receptor | |
| Antibody was raised against a peptide corresponding to 14 amino acids in the central domain of human Frizzled-6. The immunogen is located within 50-550 amino acids of Frizzled-6. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/ml | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
| PBS | |
| 8323 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title