missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-3 Antibody (169310) [DyLight 488], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1001K
This item is not returnable.
View return policy
Description
Frizzled-3 Monoclonal antibody specifically detects Frizzled-3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
Specifications
| Frizzled-3 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 169310 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| frizzled (Drosophila) homolog 3, frizzled homolog 3 (Drosophila), frizzled-3, Fz-3, hFz3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3, Met1-Val157, Accession # Q61086 | |
| 0.1 mL | |
| GPCR, Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway | |
| 7976 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction