missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24272-1-AP-20UL
This item is not returnable.
View return policy
Description
This intronless gene is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the wingless type MMTV integration site family of signaling proteins. This gene encodes a protein that is coupled to the beta-catenin canonical signaling pathway. Competition between the wingless-type MMTV integration site family, member 3A and wingless-type MMTV integration site family, member 5A gene products for binding of this protein is thought to regulate the beta-catenin-dependent and -independent pathways.Specifications
| Frizzled 2 | |
| Polyclonal | |
| Unconjugated | |
| FZD2 | |
| Frizzled 2, Fz 2, FZD2, FzE2, hFz2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2535, 57265, 64512 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q08464, Q14332, Q9JIP6 | |
| Fzd2 | |
| Frizzled 2 Fusion Protein Ag18003 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction