missing translation for 'onlineSavingsMsg'
Learn More

Frizzled-2 Antibody (2C8), Novus Biologicals™

Product Code. 18367677 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367677 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367677 Supplier Novus Biologicals Supplier No. H00002535M01100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Frizzled-2 Monoclonal antibody specifically detects Frizzled-2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frizzled-2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2C8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001457
Gene Alias frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, fz-2, Fz2, FzE2, hFz2
Host Species Mouse
Immunogen FZD2 (NP_001457, 192 a.a. ~ 245 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YATLEHPFHCPRVLKVPSYLSYKFLGERDCAAPCEPARPDGSMFFSQEETRFAR
Purification Method IgG purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, GPCR, Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2535
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.