missing translation for 'onlineSavingsMsg'
Learn More

Frizzled-2 Antibody (1E7), Novus Biologicals™

Product Code. 18382498 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382498 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382498 Supplier Novus Biologicals Supplier No. H00002535M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Frizzled-2 Monoclonal antibody specifically detects Frizzled-2 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frizzled-2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1E7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001457
Gene Alias frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, fz-2, Fz2, FzE2, hFz2
Host Species Mouse
Immunogen FZD2 (NP_001457, 192 a.a. ~ 245 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YATLEHPFHCPRVLKVPSYLSYKFLGERDCAAPCEPARPDGSMFFSQEETRFAR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, GPCR, Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2535
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.