missing translation for 'onlineSavingsMsg'
Learn More

Frizzled-10 Antibody (1023101) [Biotin], Novus Biologicals™

Product Code. 30731444 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731444 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731444 Supplier Novus Biologicals Supplier No. FAB3458B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Frizzled-10 Monoclonal antibody specifically detects Frizzled-10 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frizzled-10
Applications Immunohistochemistry
Classification Monoclonal
Clone 1023101
Conjugate Biotin
Formulation PBS
Gene Alias CD350, CD350 antigen, frizzled (Drosophila) homolog 10, frizzled homolog 10 (Drosophila), frizzled-10, Fz10, Fz-10, FzE7, hFz10
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human Frizzled-10, Ile21-Gly161, Accession # Q9ULW2
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 11211
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.