missing translation for 'onlineSavingsMsg'
Learn More

Frizzled-1 Antibody (109), DyLight 755, Novus Biologicals™

Product Code. 18376200 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376200 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376200 Supplier Novus Biologicals Supplier No. NBP290413IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Frizzled-1 Monoclonal specifically detects Frizzled-1 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frizzled-1
Applications ELISA
Classification Monoclonal
Clone 109
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias DKFZp564G072, FLJ95923, frizzled family receptor 1, frizzled homolog 1, frizzled, Drosophila, homolog of, 1, frizzled-1, fz-1, fzE1, hFz1, seven transmembrane spanning receptor, Wnt receptor
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Frizzled-1 (NP_067432.2; Met1-His248).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8321
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.