Learn More
Fritsch™ Disk Mill Pulverisette 13
Brand:  Fritsch 13.1030.00
Encompass
Product Code. 13134264
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Max feed particle size 20mm Final fineness 0.1 - 12mm, 44 x 87 x 40 cm (wxdxh) throughput up to 150kg/hr 400V 50/60Hz 1830W 3 phase Fritsch