missing translation for 'onlineSavingsMsg'
Learn More

Friedelin, TRC

Product Code. 30135443
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135443 50 mg 50mg
30134741 10 mg 10mg
30006825 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135443 Supplier Toronto Research Chemicals Supplier No. TRCF76500050MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 559-74-0
Molecular Formula C30 H50 O
Molecular Weight (g/mol) 426.72
Synonym 24,25,26-Trinoroleanan-3-one, 5,9,13-trimethyl-, (4β,5β,8α,9β,10α,13α,14β)-, D:A-Friedooleanan-3-one (8CI), Friedelin (6CI), (-)-Friedelin, (4β,5β,8α,9β,10α,13α,14β)-5,9,13-Trimethyl-24,25,26-trinoroleanan-3-one, 3(2H)-Picenone, eicosahydro-4,4a,6b,8a,11,11,12b,14a-octamethyl-, (4R,4aS,6aS,6bR,8aR,12aR,12bS,14aS,14bS)-, 3(2H)-Picenone, eicosahydro-4,4a,6b,8a,11,11,12b,14a-octamethyl-, [4R-(4α,4aα,6aβ,6bα,8aα,12aα,12bβ,14aα,14bβ)]-, 3-Oxofriedelane, Friedelan-3-one, Friedelanone, Friedeline, NSC 55141
IUPAC Name (4R,4aS,6aS,6aS,6bR,8aR,12aR,14aS,14bS)-4,4a,6a,6b,8a,11,11,14a-octamethyl-2,4,5,6,6a,7,8,9,10,12,12a,13,14,14b-tetradecahydro-1H-picen-3-one
SMILES C[C@H]1C(=O)CC[C@@H]2[C@]1(C)CC[C@H]3[C@@]2(C)CC[C@@]4(C)[C@@H]5CC(C)(C)CC[C@]5(C)CC[C@]34C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C30H50O/c1-20-21(31)9-10-22-27(20,5)12-11-23-28(22,6)16-18-30(8)24-19-25(2,3)13-14-26(24,4)15-17-29(23,30)7/h20,22-24H,9-19H2,1-8H3/t20-,22+,23-,24+,26+,27+,28-,29+,30-/m0/s1
Formula Weight 426.3862
Chemical Name or Material Friedelin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.