missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRG1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-49864
This item is not returnable.
View return policy
Description
FRG1 Polyclonal specifically detects FRG1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| FRG1 | |
| Polyclonal | |
| Western Blot, Peptide ELISA Detection limit 1:2000 | |
| NP_004468.1 | |
| FRG1 | |
| Peptide with sequence C-TVTNFGEISGT corresponding to internal region according to NP_004468.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is specific to the Internal region. Cross reacts with Human. Expected from sequence similarity: Mouse and Rat. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| facioscapulohumeral muscular dystrophy region gene-1, FSG1FRG1A, FSHD region gene 1, FSHD region gene 1 protein, protein FRG1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2483 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction