missing translation for 'onlineSavingsMsg'
Learn More

FRG1 Antibody (2B9), Novus Biologicals™

Product Code. 18352618 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352618 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352618 Supplier Novus Biologicals Supplier No. H00002483M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FRG1 Monoclonal antibody specifically detects FRG1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRG1
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 2B9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH53997
Gene Alias facioscapulohumeral muscular dystrophy region gene-1, FSG1FRG1A, FSHD region gene 1, FSHD region gene 1 protein, protein FRG1
Host Species Mouse
Immunogen FRG1 (AAH53997, 1 a.a. ~ 258 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAEYSYVKSTKLVLKGTKTKSKKKKSKDKKRKREEDEETQLDIVGIWWTVTNFGEISGTIAIEMDKGTYIHALDNGLFTLGAPHKEVDEGPSPPEQFTAVKLSDSRIALKSGYGKYLGINSDGLVVGRSDAIGPREQWEPVFQNGKMALLASNSCFIRCNEAGDIEAKSKTAGEEEMIKIRSCAERETKKKDDIPEEDKGNVKQCEINYVKKFQSFQDHKLKISKEDSKILKKARKDGFLHETLLDRRAKLKADRYCK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2483
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.