Description
- Requires castor frame or subframe
- Filtration efficiency 80%
- Mounted at the bottom of the appliance
Specifications
Specifications
| For Use With (Equipment) | UF30 Oven |
| Type | Fresh Air Filter |