Learn More
Memmert™ Fresh-Air Filter for UF260plus
Fresh-air filter designed to be mounted on the bottom of the unit. Castor frame or subframe not included.
Brand:  Memmert™ UF260plusR8
Additional Details : Weight : 0.01200kg
Product Code. 17519205
  • £249.00 / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
  • Requires castor frame or subframe
  • Filtration efficiency 80%
  • Mounted at the bottom of the appliance
Specifications
Specifications
UF260plus Oven
Fresh Air Filter