missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frequenin Mouse anti-Human, Clone: 11B8, Novus Biologicals™
Mouse Monoclonal Antibody
£250.00 - £397.00
Specifications
| Antigen | Frequenin |
|---|---|
| Clone | 11B8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222094
|
Novus Biologicals
NBP2-59480-50UL |
50 μL |
£250.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18262541
|
Novus Biologicals
NBP2-59480-100UL |
100 μL |
£397.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Frequenin Monoclonal specifically detects Frequenin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| Frequenin | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| DKFZp761L1223, FREQFLUP, frequenin (Drosophila) homolog, Frequenin homolog, frequenin homolog (Drosophila), Frequenin-like protein, Frequenin-like ubiquitous protein, NCS-1, neuronal calcium sensor 1 | |
| NCS1 | |
| IgG1 κ | |
| Protein A purified |
| 11B8 | |
| Western Blot 1:1000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 23413 | |
| Recombinant human NCS1 (1-190aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title