missing translation for 'onlineSavingsMsg'
Learn More

Frequenin Antibody (11B8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Bio-Techne NBP2-59480F

Product Code. 18249400

  • £470.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Frequenin Monoclonal antibody specifically detects Frequenin in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Frequenin
Monoclonal
FITC
DKFZp761L1223, FREQFLUP, frequenin (Drosophila) homolog, Frequenin homolog, frequenin homolog (Drosophila), Frequenin-like protein, Frequenin-like ubiquitous protein, NCS-1, neuronal calcium sensor 1
Recombinant human Frequenin (1-190aa) purified from E. coli
0.1 mL
Neuroscience
23413
Store at 4°C in the dark.
IgG1 κ
Flow Cytometry, ELISA
11B8
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.