missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FREM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13086-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizes isoform 1 (244 kDa), isoform 2 (80 kDa) and isoform 3 (43 kDa).
FREM1 is a member of the FRAS1-related extracellular matrix protein family and is thought to play a role in craniofacial and renal development. FREM1 functions as an extracellular matrix protein that is essential for epidermal adhesion during embryogenesis and may also participate in epidermal differentiation. It is recognized by cells in the embryonic skin and hair follicles through different members of the integrin family. Deficiency in the Fras1/Frem genes gives rise to the bleb phenotype, which is equivalent to the human hereditary disorder Fraser syndrome.Specifications
| FREM1 | |
| Polyclonal | |
| Unconjugated | |
| FREM1 | |
| C9orf143, C9orf145, C9orf154, FREM1, Protein QBRICK, QBRICK, RP11 439N12.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 158326 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5H8C1 | |
| FREM1 | |
| FREM1 Fusion Protein Ag3617 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction