missing translation for 'onlineSavingsMsg'
Learn More

Labconco FreeZone Stoppering Tray Dryer

Product Code. 17960180 Shop All Labconco Corporation Products
Click to view available options
Includes:
6-Port Manifold
Isolation Valve
Stoppering Tray Dryer
Unit Size:
1 unit
This item is not returnable. View return policy

Product Code. 17960180

Brand: Labconco 794801090

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

FreeZone Stoppering Tray Dryers lyophilize moderate to large loads of vials or serum bottles and stopper them under vacuum. A pneumatic mechanism stoppers containers under vacuum on three large adjustable processing shelves.

  • A pneumatic mechanism stoppers containers under vacuum on three large adjustable processing shelves, 16.5 x 12 in. each.
  • An HCFC/CFC-free refrigeration system and electric heater cool and heat fluid medium that is circulated through the shelves to maintain shelf temperatures within ±1° C.
  • Lyo-Works™ OS lets the user set up to 30 programs, with up to 16 steps each; record, chart and store data; and receive email alerts.
  • The tray dryer connects to a FreeZone Console Freeze Dryer that provides a real time display of the tray dryer’s parameters on a large capacitive touchscreen.
  • For use with any FreeZone Freeze Dryer manufactured after July 2017.

Specifications

Type Stoppering Tray Dryer
Capacity (Metric) 6 L, 12 L, or 18 L Freeze Dryer
No. of Shelves 3
Temperature Range (Metric) -40°C to +40°C
Material Stainless Steel
Includes Stoppering Tray Dryer
Plug Type All except North America
Voltage 230 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.