Learn More
Eppendorf™ Freezer Rack: Innova™ U360, U535, U725/-G, CryoCube™ F740 series (3-comp., Std),
Description
Freezer Rack: Innova™ U360, U535, U725, U725-G, CryoCube™ F740 series (3-compartment, Standard), DWP (53 mm), side access, stainless steel- Rack for DWP (53 mm/ 2 in)- Max. DWP size [mm]: 86 x 128- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 549 x 139 x 389 (21.6 x 5.5 x 15.3 in)- Max. DWP number per rack: 42 (6 x 7)- Stainless steel- For ULT freezer: Innova U360 (max. 9 racks), Innova U535 (max. 12 racks), Innova U725 and U725-G (max. 18 racks), CryoCube F740 series (3-compartment version; max. 18 racks)- For ULT freezer: Innova U360 (max. 378 DWP), Innova U535 (max. 504 DWP), Innova U725 and U725-G (max. 756 DWP), CryoCube F740 series (3-compartment version; max. 756 DWP)
Specifications
Specifications
| Product Type | Freezer Rack |