Learn More
Eppendorf™ Freezer Rack: Innova™ U360, U535, U725/-G, CryoCube™ F740 series (3-comp., Std),
Description
Freezer Rack: Innova™ U360, U535, U725, U725-G, CryoCube™ F740 series (3-compartment, Standard), 2.5 in/64 mm, side access, stainless steel- Rack for 64 mm/2.5 in boxes (5 mL tubes)- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 569 x 139 x 394 (22.4 x 5.5 x 15.5 in)- Max. box number per rack: 24 (4 x 6)- Stainless steel- For ULT freezer: Innova U360 (max. 9 racks), Innova U535 (max. 12 racks), Innova U725 and U725-G (max. 18 racks), CryoCube F740 series (3-compartment version; max. 18 racks)- For ULT freezer: Innova U360 (max. 216 boxes), Innova U535 (max. 288 boxes), Innova U725 and U725-G (max. 432 boxes), CryoCube F740 series (3-compartment version; max. 432 boxes)
Specifications
Specifications
| Product Type | Freezer Rack |