Learn More
Eppendorf™ Freezer Rack: Innova™ U101, CryoCube™ F101h, 4 in/102 mm
Product Code. 15743265
Product Code. 15743265 Supplier Eppendorf™ Supplier No. 6001061410
  • ONLINE EXCLUSIVE PRICE
    £123.00
    £105.85 / Pack of 1
    Save £17.15
    13% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ U101, CryoCube™ F101h, 4 in/102 mm, side access, stainless steel- Rack for 102 mm/4 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 290 x 142 x 214 (11.4 x 5.6 x 8.4 in)- Max. box number per rack: 4 (2 x 2)- Stainless steel- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 6 racks)- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 24 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack