Learn More
Eppendorf™ Freezer Rack: Innova™ U101, CryoCube™ F101h, 2 in/53 mm
Product Code. 15723265
Product Code. 15723265 Supplier Eppendorf™ Supplier No. 6001061210
Only null left
Add to basket
Add to basket
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ U101, CryoCube™ F101h, 2 in/53 mm, side access, stainless steel- Rack for 53 mm/2 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 290 x 142 x 279 (11.4 x 5.6 x 11 in)- Max. box number per rack: 10 (5 x 2)- Stainless steel- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 6 racks)- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 60 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack