Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, MTP
Product Code. 15713255
Product Code. 15713255
Brand: Eppendorf™ 6001040011
Only null left
Add to basket
Add to basket
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ C580, C760, locking rod, MTP, side access, stainless steel- Rack for MTP (18 mm)- Max. MTP size [mm]: 86 x 128- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 140 x 91 x 766 (5.51 x 3.58 x 30.16)- Max. MTP number per rack: 38 (38 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 52 racks), Innova C760 (max. 66 racks)- For ULT freezer: Innova C580 (max. 1,976 MTP), Innova C760 (max. 2,508 MTP)- With locking rod for increased sample safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack