Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, lock, 3 in/76 mm
Product Code. 15743225
Product Code. 15743225 Supplier Eppendorf™ Supplier No. 6001000311
  • ONLINE EXCLUSIVE PRICE
    £129.00
    £110.57 / Pack of 1
    Save £18.43
    14% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, locking rod, 3 in/76 mm, side access, stainless steel- Rack for 76 mm/3 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 148 x 141 x 726 (5.8 x 5.6 x 28.6 in)- Max. box number per rack: 9 (9 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks), CryoCube™ FC660 series (max. 36 racks)- For ULT freezer: Innova C580 (max. 288 boxes), Innova C760 (max. 360 boxes), CryoCube™ FC660 series (max. 324 boxes)- With locking rod for increased sample safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack