missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Free Chlorine Reagent Powder Pillows

Product Code. 10307182
Change view
Click to view available options
Range:
0.02 to 2.0 mg/L Cl2
0.1 to 10.0 mg/L Cl2
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10307182 0.02 to 2.0 mg/L Cl2 Each
13187429 0.1 to 10.0 mg/L Cl2 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10307182 Supplier Hach Lange™ Supplier No. 2105569

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Hach Lange Free Chlorine Reagent Powder Pillows, Range: 0.02 to 2.00 mg/L Cl2, Certifications/Compliance: EPA compliant, For Use With: Free chlorine determination, All except DR/3000, DR/2000, Parameter Specifications: Free Chlorine, Chlorine Dioxide, Quantity: 100Pack

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EPA compliant
Volume (Metric) 10 mL
Format Powder
For Use With (Application) Free chlorine determination
For Use With (Equipment) All except DR/3000, DR/2000
Parameter Specifications Free Chlorine, Chlorine Dioxide
Quantity 100 Pack
Range 0.02 to 2.0 mg/L Cl2
Storage Requirements 10°C to 25°C
Test Method 8021, DPD
Type Free Chlorine Reagent Powder Pillows
Show More Show Less
compliance-icons
Product Identifier
  • DPD Free Chlorine Reagent
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: di-Sodium hydrogen phosphate, Salt of N,N-Diethyl-p-Phenylenediamine, Ethylenedinitrilotetraacetic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.