missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ FRAX 597

Description
FRAX 597 is a potent group I PAK inhibitor (IC50 values are 8, 13 and 19 nM for PAK1, 2 and 3, respectively). FRAX 597 exhibits significant inhibition of YES1, RET, CSF1R and TEK at 100 nM, but is inactive against group II PAK isoforms (IC50 >10 μM for PAK4). FRAX 597 inhibits proliferation of pancreatic cancer and schwannoma cells in vitro and exhibits antitumor effects in mice.
Specifications
Specifications
| Chemical Name or Material | FRAX 597 |
| CAS | 1286739-19-2 |
| Synonym | 6-[2-Chloro-4-(5-thiazolyl)phenyl]-8-ethyl-2-[[4-(4-methyl-1-piperazinyl)phenyl]amino]pyrido[2,3-d]pyrimidin-7-(8H)-one |
| Quantity | 10 mg |
| Solubility | Soluble to 20mM in DMSO |
| Target | PAK Inhibitors |
| Molecular Formula | C29H28ClN7OS |
| Formula Weight | 558.1 |
| Physical Form | Solid |
| Color | Yellow |
| Show More |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?