missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frataxin Mouse anti-Human, Clone: rFXN/2124, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00
Specifications
| Antigen | Frataxin |
|---|---|
| Clone | rFXN/2124 |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA 2-4 ug/ml |
| Applications | ELISA |
Description
Frataxin Monoclonal specifically detects Frataxin in Human samples. It is validated for ELISA.Specifications
| Frataxin | |
| 0.2 mg/ml | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Diabetes Research | |
| 10 mM PBS with 0.05% BSA | |
| 2395 | |
| IgG1 κ | |
| Protein A or G purified |
| rFXN/2124 | |
| ELISA 2-4 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199 | |
| Recombinant human Frataxin protein fragment (around aa 57-210) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title