missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frataxin, Mouse anti-Human, Clone: FXN/2124, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79802-100UG
This item is not returnable.
View return policy
Description
Frataxin Monoclonal specifically detects Frataxin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Frataxin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q16595 | |
| FXN | |
| Recombinant fragment of human Frataxin protein (around aa 57-210) (exact sequence is proprietary) | |
| 100 μg | |
| Diabetes Research | |
| 2395 | |
| Store at 4°C. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| FXN/2124 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction