missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frataxin Antibody (rFXN/2124), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08576FR
This item is not returnable.
View return policy
Description
Frataxin Monoclonal antibody specifically detects Frataxin in Human samples. It is validated for ELISA
Specifications
| Frataxin | |
| Monoclonal | |
| DyLight 680 | |
| CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199 | |
| Recombinant human Frataxin protein fragment (around aa 57-210) (exact sequence is proprietary) (Uniprot: Q16595) | |
| 100 μg | |
| Diabetes Research | |
| 2395 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| rFXN/2124 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction