missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frataxin Antibody (OTI3C7), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70769PCP
This item is not returnable.
View return policy
Description
Frataxin Monoclonal antibody specifically detects Frataxin in Human samples. It is validated for Western Blot
Specifications
| Frataxin | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| OTI3C7 | |
| Western Blot | |
| CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199 | |
| Full length human recombinant protein of human FXN(NP_000135) produced in HEK293T cell. | |
| 0.1 mL | |
| Diabetes Research | |
| 2395 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction