missing translation for 'onlineSavingsMsg'
Learn More

Frataxin Antibody (FXN/2124), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-79936AF700

Product Code. 18260337

  • £466.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Frataxin Monoclonal specifically detects Frataxin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Frataxin
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
FXN
Recombinant fragment of human Frataxin (FXN) protein (around aa 57-210) (exact sequence is proprietary) (Uniprot: Q16595)
0.1 mL
Diabetes Research
Human
IgG2b κ
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
FXN/2124
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.