missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRAT3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1008
This item is not returnable.
View return policy
Description
FRAT3 Polyclonal specifically detects FRAT3 in Mouse samples. It is validated for Peptide ELISA.
Specifications
| FRAT3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| Frat3, frequently rearranged in advanced T-cell lymphomas 3 Official Gene Symbol - Peg12GenBank Accession Number NP_038816 LocusLink ID - 27412 (mouse), paternally expressed 12, Principal Names -Peg12 | |
| Peptide with sequence C-SLAANNPGGHSTNL corresponding to C-Terminus according to NP_038816.1. | |
| 0.1 mg | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:2000 | |
| NP_038816. | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction